Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TENM4
Entrez Gene ID
395406
Gene Name
teneurin transmembrane protein 4
Gene Aliases
ODZ4, TEN4
Chromosome Location
Chr.1: 193208187 - 193804627 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001354731.2 NP_001341660.1 4 - 5 846 99
GenBank mRNA BX932994.1 - 3 - 4 603 99

View Full Product Details