Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PBLD
Entrez Gene ID
423685
Gene Name
phenazine biosynthesis like protein domain containing
Gene Aliases
-
Chromosome Location
Chr.6: 11263100 - 11281943 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277337.1 NP_001264266.1 6 - 7 705 56
XM_015278680.2 XP_015134166.1 6 - 7 564 56
GenBank mRNA BX934609.1 - 6 - 7 583 56

View Full Product Details