Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP6AP1L
Entrez Gene ID
427324
Gene Name
ATPase H+ transporting accessory protein 1 like
Gene Aliases
-
Chromosome Location
Chr.Z: 63542036 - 63562670 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA CR352983.1 - 3 - 4 318 102

View Full Product Details