The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252285.1 | NP_001239214.1 | 3 - 4 | 496 | 100 |
GenBank mRNA | CR353823.1 | - | 2 - 3 | 289 | 100 |