Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP3A4
Entrez Gene ID
416477
Gene Name
cytochrome P450 family 3 subfamily A member 4
Gene Aliases
CYP3A80
Chromosome Location
Chr.14: 4288049 - 4299525 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001329508.1 NP_001316437.1 10 - 11 1088 84
XM_015294357.2 XP_015149843.1 10 - 11 1139 84
GenBank mRNA BX931257.1 - 10 - 11 1089 84

View Full Product Details