Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MRPL15
Entrez Gene ID
421121
Gene Name
mitochondrial ribosomal protein L15
Gene Aliases
-
Chromosome Location
Chr.2: 110380995 - 110389411 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006388.1 NP_001006388.1 2 - 3 309 97
GenBank mRNA AJ719996.1 - 2 - 3 309 97

View Full Product Details