Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SNCB
Entrez Gene ID
395391
Gene Name
synuclein beta
Gene Aliases
-
Chromosome Location
Chr.13: 11057144 - 11060856 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204671.1 NP_990002.1 4 - 5 413 109
XM_015293731.2 XP_015149217.1 4 - 5 451 109
GenBank mRNA AF253514.1 - 4 - 5 413 109

View Full Product Details