Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
WNT3A
Entrez Gene ID
395396
Gene Name
Wnt family member 3A
Gene Aliases
WNT-3A
Chromosome Location
Chr.2: 2551903 - 2634749 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001171601.1 NP_001165072.1 2 - 3 503 66
NM_204675.2 NP_990006.2 2 - 3 401 66
GenBank mRNA AB024080.1 - 2 - 3 401 66
CR386781.1 - 1 - 2 478 66
DQ022307.1 - 2 - 3 506 66
EF068232.1 - 2 - 3 331 66

View Full Product Details