Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB2A
Entrez Gene ID
396153
Gene Name
RAB2A, member RAS oncogene family
Gene Aliases
RAB2
Chromosome Location
Chr.2: 112818651 - 112859453 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205228.1 NP_990559.1 7 - 8 540 92
GenBank mRNA X84220.1 - - 540 92

View Full Product Details