Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LHX1
Entrez Gene ID
396381
Gene Name
LIM homeobox 1
Gene Aliases
LIM-1
Chromosome Location
Chr.19: 8536207 - 8540685 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205413.1 NP_990744.1 2 - 3 539 117
XM_015295682.2 XP_015151168.1 2 - 3 524 117
GenBank mRNA L35569.1 - 2 - 3 539 117

View Full Product Details