Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
GATA6
Entrez Gene ID
396390
Gene Name
GATA binding protein 6
Gene Aliases
GATA-6
Chromosome Location
Chr.2: 102636810 - 102654409 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205420.1 NP_990751.1 5 - 6 998 63
XM_015277936.2 XP_015133422.2 6 - 7 1405 63
GenBank mRNA U11889.1 - 5 - 6 998 63

View Full Product Details