Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CAPZA1
Entrez Gene ID
396521
Gene Name
capping actin protein of muscle Z-line alpha subunit 1
Gene Aliases
-
Chromosome Location
Chr.26: 3533331 - 3543277 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205515.1 NP_990846.1 - 536 99
GenBank mRNA AJ719691.1 - - 226 99
M25534.1 - - 536 99

View Full Product Details