Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RAB18
Entrez Gene ID
420483
Gene Name
RAB18, member RAS oncogene family
Gene Aliases
-
Chromosome Location
Chr.2: 15786145 - 15799223 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006355.1 NP_001006355.1 3 - 4 248 121
GenBank mRNA AJ719773.1 - 3 - 4 248 121

View Full Product Details