Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
FGF8
Entrez Gene ID
396313
Gene Name
fibroblast growth factor 8
Gene Aliases
FGF-8
Chromosome Location
Chr.6: 23669271 - 23677126 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012767.1 NP_001012785.1 1 - 2 33 56
XM_015288595.2 XP_015144081.1 1 - 2 248 56
GenBank mRNA FJ882410.1 - 1 - 2 33 56
U55189.1 - 1 - 2 33 56

View Full Product Details