Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CEL
Entrez Gene ID
417165
Gene Name
carboxyl ester lipase
Gene Aliases
-
Chromosome Location
Chr.17: 7073101 - 7078463 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012997.1 NP_001013015.1 1 - 2 83 105
GenBank mRNA BX931584.2 - 1 - 2 71 105
BX931585.2 - 1 - 2 81 105
BX931587.2 - 1 - 2 83 105
BX933612.1 - 1 - 2 83 105
BX950478.1 - 1 - 2 83 105

View Full Product Details