Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ARPC1B
Entrez Gene ID
416490
Gene Name
actin related protein 2/3 complex subunit 1B
Gene Aliases
-
Chromosome Location
Chr.14: 4766374 - 4773039 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030632.1 NP_001025803.1 5 - 6 646 60
XM_015294172.2 XP_015149658.1 5 - 6 679 60
GenBank mRNA AJ720885.1 - 5 - 6 646 60

View Full Product Details