Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NSFL1C
Entrez Gene ID
419268
Gene Name
NSFL1 cofactor
Gene Aliases
-
Chromosome Location
Chr.20: 9807241 - 9814899 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030859.1 NP_001026030.1 2 - 3 209 134
XM_015296393.2 XP_015151879.1 2 - 3 439 134
GenBank mRNA AJ720274.1 - 2 - 3 209 134

View Full Product Details