Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SAMD11
Entrez Gene ID
419434
Gene Name
sterile alpha motif domain containing 11
Gene Aliases
-
Chromosome Location
Chr.21: 2921607 - 3010295 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030877.1 NP_001026048.1 2 - 3 264 94
XM_015296882.2 XP_015152368.2 4 - 5 1072 94
XM_015296883.2 XP_015152369.2 4 - 5 1021 94
XM_015296886.2 XP_015152372.1 3 - 4 414 94
XM_015296887.2 XP_015152373.1 3 - 4 242 94
XM_025142516.1 XP_024998284.1 3 - 4 1162 94
XM_025142517.1 XP_024998285.1 8 - 9 1521 94
XM_025142518.1 XP_024998286.1 2 - 3 965 94
GenBank mRNA AJ720450.1 - 2 - 3 264 94

View Full Product Details