Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CYP7A1
Entrez Gene ID
414834
Gene Name
cytochrome P450 family 7 subfamily A member 1
Gene Aliases
-
Chromosome Location
Chr.2: 112027114 - 112034638 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001001753.1 NP_001001753.1 2 - 3 356 81
GenBank mRNA AB109636.1 - 2 - 3 356 81

View Full Product Details