Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
LAPTM4A
Entrez Gene ID
421959
Gene Name
lysosomal protein transmembrane 4 alpha
Gene Aliases
-
Chromosome Location
Chr.3: 101413593 - 101426398 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031092.1 NP_001026263.1 4 - 5 491 102
GenBank mRNA AJ719367.1 - 4 - 5 491 102
BX950470.1 - 4 - 5 441 102

View Full Product Details