Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RFFL
Entrez Gene ID
417529
Gene Name
ring finger and FYVE like domain containing E3 ubiquitin protein ligase
Gene Aliases
-
Chromosome Location
Chr.19: 4627665 - 4651321 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030711.1 NP_001025882.1 3 - 4 617 65
XM_015295722.2 XP_015151208.1 3 - 4 622 65
XM_015295723.2 XP_015151209.1 3 - 4 682 65
XM_015295725.2 XP_015151211.1 3 - 4 693 65
XM_025141948.1 XP_024997716.1 4 - 5 1907 65
GenBank mRNA AJ720958.1 - 3 - 4 617 65

View Full Product Details