Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PARPBP
Entrez Gene ID
418092
Gene Name
PARP1 binding protein
Gene Aliases
C12orf48, C1H12ORF48, PARI
Chromosome Location
Chr.1: 55369758 - 55417957 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030763.2 NP_001025934.2 5 - 6 760 75
XM_025148736.1 XP_025004504.1 5 - 6 753 75
XM_025148737.1 XP_025004505.1 5 - 6 763 75
GenBank mRNA AJ720065.1 - 5 - 6 760 75

View Full Product Details