Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
COPS7A
Entrez Gene ID
418283
Gene Name
COP9 signalosome subunit 7A
Gene Aliases
-
Chromosome Location
Chr.1: 77817352 - 77820482 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030775.1 NP_001025946.1 6 - 7 891 97
GenBank mRNA AJ720533.1 - 6 - 7 891 97
CR389600.1 - 6 - 7 722 97

View Full Product Details