Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHMP2B
Entrez Gene ID
418461
Gene Name
charged multivesicular body protein 2B
Gene Aliases
-
Chromosome Location
Chr.1: 94275896 - 94287427 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030792.2 NP_001025963.1 4 - 5 475 113
GenBank mRNA AJ851748.1 - 4 - 5 476 113

View Full Product Details