Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DCUN1D1
Entrez Gene ID
424963
Gene Name
defective in cullin neddylation 1 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.9: 16569451 - 16592139 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031318.1 NP_001026489.1 2 - 3 297 133
GenBank mRNA AJ719993.1 - 2 - 3 297 133

View Full Product Details