Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AK6
Entrez Gene ID
425215
Gene Name
adenylate kinase 6
Gene Aliases
-
Chromosome Location
Chr.Z: 64365002 - 64369042 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031333.1 NP_001026504.1 2 - 3 174 109
GenBank mRNA AJ851392.1 - 2 - 3 174 109

View Full Product Details