Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAM76A
Entrez Gene ID
426384
Gene Name
family with sequence similarity 76 member A
Gene Aliases
-
Chromosome Location
Chr.23: 2233904 - 2249335 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031377.2 NP_001026548.1 4 - 5 392 85
GenBank mRNA AJ720569.1 - 4 - 5 371 85
CR386761.1 - 4 - 5 403 85

View Full Product Details