Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
COLEC10
Entrez Gene ID
420281
Gene Name
collectin subfamily member 10
Gene Aliases
CL-1
Chromosome Location
Chr.2: 136385334 - 136405870 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039598.1 NP_001034687.1 2 - 3 252 78
XM_015283021.2 XP_015138507.1 2 - 3 352 78
GenBank mRNA DQ129668.1 - 2 - 3 252 78

View Full Product Details