Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PNO1
Entrez Gene ID
769547
Gene Name
partner of NOB1 homolog
Gene Aliases
-
Chromosome Location
Chr.3: 11863403 - 11866951 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079490.1 NP_001072958.1 4 - 5 564 101
GenBank mRNA AJ851486.1 - 4 - 5 564 101

View Full Product Details