Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RRH
Entrez Gene ID
771805
Gene Name
retinal pigment epithelium-derived rhodopsin homolog
Gene Aliases
-
Chromosome Location
Chr.4: 58101325 - 58112439 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001079759.2 NP_001073227.1 2 - 3 402 84
GenBank mRNA AY339626.1 - 2 - 3 406 84

View Full Product Details