Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NLGN1
Entrez Gene ID
424980
Gene Name
neuroligin 1
Gene Aliases
neuroligin-1
Chromosome Location
Chr.9: 18877811 - 19231069 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081502.1 NP_001074971.1 5 - 6 982 71
XM_015291652.2 XP_015147138.1 6 - 7 1908 71
XM_015291653.2 XP_015147139.1 6 - 7 1954 71
XM_015291654.2 XP_015147140.1 6 - 7 1450 71
XM_015291656.2 XP_015147142.1 5 - 6 1847 71
XM_015291659.2 XP_015147145.1 4 - 5 1787 71
GenBank mRNA EF199811.1 - 5 - 6 982 71
GQ489204.1 - 5 - 6 982 71
GU080338.1 - 4 - 5 922 71
GU080339.1 - 4 - 5 922 71
GU080340.1 - 3 - 4 862 71
GU574580.1 - 5 - 6 982 71

View Full Product Details