Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CASP14
Entrez Gene ID
776274
Gene Name
caspase 14, apoptosis-related cysteine peptidase
Gene Aliases
CASP17
Chromosome Location
Chr.22: 5316342 - 5320143 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113293.1 NP_001106764.1 4 - 5 562 80
XM_015272947.2 XP_015128433.1 3 - 4 1073 80
GenBank mRNA EU165361.1 - 3 - 4 504 80
EU165362.1 - 4 - 5 562 80

View Full Product Details