The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_204182.2 | NP_989513.2 | 1 - 2 | 207 | 145 |
GenBank mRNA | CO421284.1 | - | 1 - 2 | 215 | 145 |