Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SMN
Entrez Gene ID
374025
Gene Name
survival of motor neuron
Gene Aliases
-
Chromosome Location
Chr.Z: 73237466 - 73242999 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204199.1 NP_989530.1 3 - 4 237 83
XM_015280581.2 XP_015136067.1 3 - 4 259 83
XM_015280582.2 XP_015136068.1 3 - 4 259 83
GenBank mRNA AF322650.1 - 3 - 4 237 83

View Full Product Details