Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ATP4B
Entrez Gene ID
386581
Gene Name
ATPase H+/K+ transporting beta subunit
Gene Aliases
-
Chromosome Location
Chr.1: 138638385 - 138644098 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_204418.1 NP_989749.1 5 - 6 761 82
GenBank mRNA JQ277718.1 - 1 - 2 49 82
L08047.1 - 5 - 6 761 82

View Full Product Details