Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DHODH
Entrez Gene ID
415876
Gene Name
dihydroorotate dehydrogenase (quinone)
Gene Aliases
-
Chromosome Location
Chr.11: 19719049 - 19722544 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006131.2 NP_001006131.1 6 - 7 1054 59
GenBank mRNA AJ721004.1 - 6 - 7 1055 59

View Full Product Details