Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
NELFB
Entrez Gene ID
417254
Gene Name
negative elongation factor complex member B
Gene Aliases
COBRA1
Chromosome Location
Chr.17: 2266800 - 2277936 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006202.1 NP_001006202.1 5 - 6 812 90
GenBank mRNA AJ719725.1 - 5 - 6 812 90

View Full Product Details