Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC16A1
Entrez Gene ID
419875
Gene Name
solute carrier family 16 member 1
Gene Aliases
-
Chromosome Location
Chr.26: 3611402 - 3628787 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006323.1 NP_001006323.1 - 287 66
XM_015298930.2 XP_015154416.1 - 283 66
XM_025143692.1 XP_024999460.1 - 1811 66
GenBank mRNA AJ719588.1 - - 287 66
AJ721138.1 - - 253 66

View Full Product Details