Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLAU
Entrez Gene ID
396424
Gene Name
plasminogen activator, urokinase
Gene Aliases
-
Chromosome Location
Chr.6: 16341341 - 16349514 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205443.2 NP_990774.2 3 - 4 204 105
GenBank mRNA J05187.1 - 3 - 4 204 105

View Full Product Details