Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IFNGR1
Entrez Gene ID
421685
Gene Name
interferon gamma receptor 1
Gene Aliases
-
Chromosome Location
Chr.3: 54895422 - 54914165 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130387.1 NP_001123859.1 4 - 5 591 114
GenBank mRNA EU057149.1 - 4 - 5 591 114

View Full Product Details