Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
OPN5
Entrez Gene ID
428670
Gene Name
opsin 5
Gene Aliases
GPR136, GRP136, Opn5m, PGR12, TMEM13, cOpn5m, neuropsin, opsin-5
Chromosome Location
Chr.3: 110092843 - 110119982 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130743.1 NP_001124215.1 1 - 2 68 96
GenBank mRNA AB368182.1 - 1 - 2 68 96

View Full Product Details