Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MGD1
Entrez Gene ID
547469
Gene Name
monogalactosyldiacylglycerol synthase 1
Gene Aliases
GmMGD1
Chromosome Location
Chr.14: 10842103 - 10847040 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250001.2 NP_001236930.1 4 - 5 1147 103
GenBank mRNA AB047475.1 - 4 - 5 1148 103

View Full Product Details