Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC548033
Entrez Gene ID
548033
Gene Name
beta-ketoacyl-acyl carrier protein synthase III
Gene Aliases
KAS III
Chromosome Location
Chr.18: 54072829 - 54077117 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250806.1 NP_001237735.1 3 - 4 590 67
GenBank mRNA AF260565.1 - 3 - 4 590 67

View Full Product Details