Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SBP
Entrez Gene ID
606297
Gene Name
selenium binding protein
Gene Aliases
-
Chromosome Location
Chr.8: 8307249 - 8310573 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AJ401230.1 - 3 - 4 540 78

View Full Product Details