Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
EN1
Entrez Gene ID
100101913
Gene Name
endonuclease
Gene Aliases
-
Chromosome Location
Chr.15: 5202032 - 5204030 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001248396.1 NP_001235325.1 1 - 2 137 98
GenBank mRNA AK246074.1 - 1 - 2 178 98
BT097673.1 - 1 - 2 175 98
EF521872.1 - 1 - 2 137 98

View Full Product Details