Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
WRKY48
Entrez Gene ID
100127392
Gene Name
WRKY48 protein
Gene Aliases
-
Chromosome Location
Chr.8: 9137643 - 9150837 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250292.1 NP_001237221.1 - 693 85
GenBank mRNA EU019584.1 - - 693 85

View Full Product Details