Gene Transcripts

Gene Symbol
BCL7A
Entrez Gene ID
605
Gene Name
BCL tumor suppressor 7A
Gene Aliases
BCL7
Chromosome Location
Chr. 12: 122021886 - 122062044 on Build GRCh38
UniGene ID
Hs.530970
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001024808.2 NP_001019979.1 2, 3 369
NM_020993.4 NP_066273.1 2, 3 369
GenBank mRNA AK296159.1 - 2, 3 371
AK316486.1 - 2, 3 267
BC094723.1 - 2, 3 180 5300018
BC113627.1 - 2, 3 193 8322679
BM801030.1 - 328
X89984.1 - 2, 3 1116

View Full Product Details