Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000740.2 | NP_000731.1 | 5 | 1718 | |
XM_005273032.3 | XP_005273089.1 | 6 | 2023 | ||
XM_011544041.2 | XP_011542343.1 | 7 | 2122 | ||
XM_011544043.2 | XP_011542345.1 | 9 | 2455 | ||
XM_011544044.2 | XP_011542346.1 | 8 | 2226 | ||
XM_011544045.2 | XP_011542347.1 | 8 | 2351 | ||
XM_011544046.2 | XP_011542348.1 | 6 | 2059 | ||
XM_011544047.2 | XP_011542349.1 | 6 | 1995 | ||
XM_017000152.1 | XP_016855641.1 | 8 | 5048 | ||
XM_017000153.1 | XP_016855642.1 | 7 | 2126 | ||
XM_017000154.1 | XP_016855643.1 | 6 | 1820 | ||
XM_017000155.1 | XP_016855644.1 | 7 | 2163 | ||
XM_017000156.1 | XP_016855645.1 | 7 | 2127 | ||
XM_017000157.1 | XP_016855646.1 | 5 | 1728 | ||
XM_017000158.1 | XP_016855647.1 | 5 | 1960 | ||
XM_017000159.1 | XP_016855648.1 | 5 | 1932 | ||
XM_017000160.1 | XP_016855649.1 | 4 | 2782 | ||
XM_017000161.1 | XP_016855650.1 | 6 | 2064 | ||
XM_017000162.1 | XP_016855651.1 | 5 | 1896 | ||
XM_017000163.1 | XP_016855652.1 | 4 | 5758 | ||
GenBank mRNA | AF279779.1 | - | 5 | 1717 | |
AF385589.1 | - | 1 | 939 | ||
AF498917.1 | - | 1 | 939 | ||
AK074575.1 | - | 1 | 1096 | ||
BC096844.1 | - | 1 | 940 | ||
BC121026.2 | - | 2 | 1082 | 40117429 | |
BC121027.2 | - | 1 | 904 |