Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004075.4 | NP_004066.1 | 8 | 2053 | |
XM_017018832.1 | XP_016874321.1 | 9 | 2188 | ||
GenBank mRNA | AK098615.1 | - | 3 | 456 | |
AK125915.1 | - | 10 | 1716 | ||
AK290552.1 | - | 8 | 1353 | ||
BC017466.1 | - | 8 | 1476 | ||
BC030519.1 | - | 8 | 1726 | 5169081 | |
D83702.1 | - | 8 | 1743 | ||
D84657.1 | - | 8 | 1157 |