Gene Transcripts

Gene Symbol
CLN8
Entrez Gene ID
2055
Gene Name
ceroid-lipofuscinosis, neuronal 8
Gene Aliases
C8orf61, EPMR
Chromosome Location
Chr. 8: 1755778 - 1786570 on Build GRCh38
UniGene ID
Hs.127675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018941.3 NP_061764.2 3 1022
XM_005266021.3 XP_005266078.1 3 966
XM_005266022.1 XP_005266079.1 3 1145
XM_005266023.1 XP_005266080.1 3 863
XM_011534745.1 XP_011533047.1 3 967
XM_011534746.2 XP_011533048.1 3 864
GenBank mRNA AF123757.1 - 3 951
AF123758.1 - 3 951
AF123759.1 - 3 951
AF123760.1 - 3 951
AF123761.1 - 3 951
AK312486.1 - 3 965
BC007725.2 - 3 876 4303103
BT007049.1 - 2 717

View Full Product Details